3,5-Dimethoxyaniline ≥98%
Supplier: BeanTown Chemical
CAS: 10272-07-8; EC No: 233-616-5; MDL No: MFCD00008392
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Powder/Needles; Molecular Formula: C8H11NO2; MW: 153.18
Melting Point: 54-57°; Boiling Point: 178°/20 mmHg; Flash point: 135°C (275°F)
- Powder/Needles
Formula:
C₈H₁₁NO₂ Boiling Pt: 178 °C/20 mmHg Melting Pt: 54…57 °C Flash Pt: 135 °C (275 °F) |
MDL Number:
MFCD00008392 CAS Number: 10272-07-8 EINECS: 233-616-5 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...