Chromium(III) fluoride tetrahydrate

Supplier: BeanTown Chemical

Synonyms: Chromium trifluoride tetrahydrate, Chromic fluoride tetrahydrate, Chromium(III) fluoride hydrate

136550-1KG 136550-250G
BT136550-1KGEA 88 USD
BT136550-1KG BT136550-250G
Chromium(III) fluoride tetrahydrate
Chromium(III) fluoride tetrahydrate

CAS: 123333-98-2; EC No: 232-137-9; MDL No: MFCD00016045 UN No: UN1756; Haz Class: 8; Packing Group: II Crystalline; Molecular Formula: CrF3·xH2O; MW: 108.99 (anhydrous) Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: CrF₃·4H₂O
MW: 181.05
MDL Number: MFCD00016045
CAS Number: 123333-98-2
EINECS: 232-137-9
UN: 1756
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR