Chlorpheniramine maleate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Chlorphenamine maleate
136540-25G
136540-5G
BT136540-25GEA
83
USD
BT136540-25G
BT136540-5G
Chlorpheniramine maleate ≥99%
Chlorpheniramine maleate
CAS: 113-92-8; EC No: 205-054-0; MDL No: MFCD00069225; RTECS: US6504000
UN No: UN2811 ; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C16H19ClN2·C4H4O4; MW: 390.86
Melting Point: 130-135°
- RTECS: US6504000
- Solid
Formula:
C₁₆H₁₉ClN₂ MW: 390.86 g/mol Melting Pt: 130…135 °C |
MDL Number:
MFCD00069225 CAS Number: 113-92-8 EINECS: 205-054-0 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...