(±)-Epichlorhydrin ≥99%

Supplier: BeanTown Chemical

Synonyms: (±)-1-Chloro-2,3-epoxypropane, Epichlorohydrin, (±)-3-Chloro-1,2-epoxypropane, 1-Chloro-2,3-epoxypropane, Epichlorhydrin, (±)-Glycerol epichlorhydrin, 3-Chloro-1,2-epoxypropane

136530-100G 136530-2.5KG 136530-500G
BT136530-100GEA 27.95 USD
BT136530-100G BT136530-2.5KG BT136530-500G
(±)-Epichlorhydrin ≥99%
(±)-Epichlorhydrin

CAS: 106-89-8; EC No: 203-439-8; MDL No: MFCD00005132; RTECS: TX4900000 UN No: UN2023; Haz Class: 6.1 (3); Packing Group: II Liquid; Molecular Formula: C3H5ClO; MW: 92.53 Boiling Point: 114-117°; Flash point: 32°C (90°F) Density (g/mL): 1.182; Refractive Index: 1.438


  • RTECS: TX4900000
  • Refractive index: 1.438
  • Liquid

Formula: C₃H₅ClO
MW: 92.52 g/mol
MDL Number: MFCD00005132
CAS Number: 106-89-8

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR