Palladium ≥99.9% (trace metals basis), powder -60 mesh
Supplier: BeanTown Chemical
CAS: 7440-05-3; EC No: 231-115-6; MDL No: MFCD00011167; RTECS: RT3480500
UN No: UN3089; Haz Class: 4.1; Packing Group: III
Powder; Molecular Formula: Pd; MW: 106.42
Melting Point: 1554°; Boiling Point: 2970°
Density (g/mL): 12.02
- RTECS: RT3480500
- Powder
Formula:
Pd Boiling Pt: 2970 °C Melting Pt: 1554 °C Density: 12.02 g/mL |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 EINECS: 231-115-6 ADR: 4.1,III |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...