Ethyl propenyl ether cis- and trans-mixture ≥97%

Supplier: BeanTown Chemical

Synonyms: Ethyl 1-Propenyl ether

136515-100G 136515-25G 136515-5G
BT136515-100GEA 251 USD
BT136515-100G BT136515-25G BT136515-5G
Ethyl propenyl ether cis- and trans-mixture ≥97%
Ethyl propenyl ether

CAS: 928-55-2; EC No: 213-176-0; MDL No: MFCD00015184; RTECS: KO0525000 UN No: UN3271; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3CH=CHOCH2CH3 ; Molecular Formula: C5H10O; MW: 86.13 Boiling Point: 67-76°; Flash point: 20°C (68°F) Density (g/mL): 0.778; Refractive Index: 1.398


  • RTECS: KO0525000
  • Refractive index: 1.398
  • Liquid

Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 67…76 °C
Density: 0.778 g/mL
Flash Pt: 20 °C (68 °F)
MDL Number: MFCD00015184
CAS Number: 928-55-2
EINECS: 213-176-0
UN: 3271
ADR: 3 II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR