Iridium(III) chloride, anhydrous
Supplier: BeanTown Chemical
136495-1G
136495-5G
BT136495-1GEA
357.33
USD
BT136495-1G
BT136495-5G
Iridium(III) chloride, anhydrous
Iridium(III) chloride
CAS: 10025-83-9; EC No: 233-044-6; MDL No: MFCD00011063
UN No: UN3260; Haz Class: 8; Packing Group: II
Powder; Molecular Formula: IrCl3; MW: 298.56
Melting Point: 763°
Hygroscopic
- Hygroscopic
- Powder
Formula:
Cl₃Ir Melting Pt: 763 °C |
MDL Number:
MFCD00011063 CAS Number: 10025-83-9 EINECS: 233-044-6 UN: 3260 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...