1,4-Bis(trimethylsilyl)-1,3-butadiyne ≥98%
Supplier: BeanTown Chemical
CAS: 4526-07-2; MDL No: MFCD00009839
UN No: UN1325; Haz Class: 4.1; Packing Group: II
Powder/Flakes; Linear Formula: (CH3)3Si-C≡C-C≡C-Si(CH3)3; Molecular Formula: C10H18Si2; MW: 194.42
Melting Point: 111-113°
Density (g/mL): 0.974; Refractive Index: 1.384
- Refractive index: 1.384
- Powder/Flakes
Formula:
C₁₀H₁₈Si₂ Melting Pt: 111…113 °C Density: 0.974 g/mL |
MDL Number:
MFCD00009839 CAS Number: 4526-07-2 UN: 1325 ADR: 4.1,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...