4-tert-Butyl-α-chlorotoluene ≥99%

Supplier: BeanTown Chemical

Synonyms: p-tert-Butylbenzyl chloride, 4-tert-Butylbenzyl chloride, p-tert-Butyl-α-chlorotoluene, 1-(tert-Butyl)-4-(chloromethyl)benzene

136480-25G 136480-5G
BT136480-25GEA 219 USD
BT136480-25G BT136480-5G
4-tert-Butyl-α-chlorotoluene ≥99%
4-tert-Butyl-α-chlorotoluene

CAS: 19692-45-6; EC No: 243-228-8; MDL No: MFCD00000918 UN No: UN1760; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C11H15Cl ; MW: 182.69 Boiling Point: 101-105°/7 mmHg; Flash point: 94°C (201°F) Density (g/mL): 0.945; Refractive Index: 1.521


  • Refractive index: 1.521
  • Liquid

Formula: C₁₁H₁₅Cl
MW: 182.69 g/mol
Boiling Pt: 101…105 °C/7 mmHg
Density: 0.945 g/mL
Flash Pt: 94 °C (201 °F)
MDL Number: MFCD00000918
CAS Number: 19692-45-6
EINECS: 243-228-8
UN: 1760
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR