Oxalyl dichloride ≥98%

Supplier: BeanTown Chemical

Synonyms: Oxalyl chloride

136460-250G 136460-50G
BT136460-250GEA 151.41 USD
BT136460-250G BT136460-50G
Oxalyl dichloride ≥98%
Oxalyl dichloride

CAS: 79-37-8; EC No: 201-200-2; MDL No: MFCD00000704; RTECS: KI2950000 UN No: UN2927; Haz Class: 6.1 (8); Packing Group: II Liquid; Linear Formula: ClCOCOCl; Molecular Formula: C2Cl2O2; MW: 126.93 Melting Point: -10 - -8°; Boiling Point: 62-65° Density (g/mL): 1.455; Refractive Index: 1.429 Moisture Sensitive


  • RTECS: KI2950000
  • Refractive index: 1.429
  • Moisture Sensitive
  • Liquid

Formula: C₂Cl₂O₂
MW: 126.93 g/mol
Boiling Pt: 62…65 °C
Melting Pt: –10 – –8 °C
Density: 1.455 g/mL
MDL Number: MFCD00000704
CAS Number: 79-37-8
EINECS: 201-200-2
UN: 2927
ADR: 4.3,I

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR