4-Methylmorpholine ≥99%

Supplier: BeanTown Chemical

Synonyms: NMM, N-Methylmorpholine

136455-100ML 136455-2.5L 136455-500ML
BT136455-100MLEA 33.4 USD
BT136455-100ML BT136455-2.5L BT136455-500ML
4-Methylmorpholine ≥99%
4-Methylmorpholine

CAS: 109-02-4; EC No: 203-640-0; MDL No: MFCD00006175; RTECS: QE5775000 UN No: UN2535; Haz Class: 3 (8); Packing Group: II Liquid; Molecular Formula: C5H11NO; MW: 101.15 Melting Point: -66°; Boiling Point: 115-116°/750 mmHg; Flash point: 14°C (57°F) Density (g/mL): 0.92; Refractive Index: 1.435


  • RTECS: QE5775000
  • Refractive index: 1.435
  • Liquid

Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 115…116 °C/750 mmHg
Melting Pt: –66 °C
Density: 0.92 g/mL
Flash Pt: 14 °C (57 °F)
MDL Number: MFCD00006175
CAS Number: 109-02-4
EINECS: 203-640-0
UN: 2535
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR