Acetyl bromide ≥98%

Supplier: BeanTown Chemical

136405-100G 136405-25G 136405-500G
BT136405-100GEA 65.39 USD
BT136405-100G BT136405-25G BT136405-500G
Acetyl bromide ≥98%
Acetyl bromide

CAS: 506-96-7; EC No: 208-061-7; MDL No: MFCD00000114; RTECS: AO5955000 UN No: UN1716; Haz Class: 8; Packing Group: II Liquid; Linear Formula: CH3COBr; MW: 122.95 Melting Point: -96°; Boiling Point: 75-77°; Flash point: 110°C (230°F) Density (g/mL): 1.663; Refractive Index: 1.45 Moisture Sensitive


  • RTECS: AO5955000
  • Refractive index: 1.45
  • Moisture Sensitive
  • Liquid

Formula: C₂H₃BrO
MW: 122.95 g/mol
Boiling Pt: 75…77 °C
Melting Pt: –96 °C
Density: 1.663 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00000114
CAS Number: 506-96-7
EINECS: 208-061-7
UN: 1716
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR