Gallium trinitrate 9 - 10% (w/w) in water
Supplier: BeanTown Chemical
136375-250G
136375-50G
BT136375-250GEA
462
USD
BT136375-250G
BT136375-50G
Gallium trinitrate 9 - 10% (w/w) in water
Gallium trinitrate
CAS: 13494-90-1; EC No: 236-815-5; MDL No: MFCD00011016
UN No: UN3218; Haz Class: 5.1; Packing Group: III
Liquid; Linear Formula: Ga(NO3)3 ; MW: 255.73
- Liquid
Formula:
GaN₃O₉ |
MDL Number:
MFCD00011016 CAS Number: 13494-90-1 EINECS: 236-815-5 ADR: 5.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...