Tin(II) acetate ≥95%
Supplier: BeanTown Chemical
Synonyms:
Tin(II) Acetate
136355-25G
136355-5G
BT136355-25GEA
223
USD
BT136355-25G
BT136355-5G
Tin(II) acetate ≥95%
Tin(II) acetate
CAS: 638-39-1; EC No: 211-335-9; MDL No: MFCD00054408
UN No: UN3146; Haz Class: 6.1; Packing Group: III
Powder/Lump; Linear Formula: Sn(OOCCH3)2; MW: 236.80
Melting Point: 180-182°
Density (g/mL): 2.31
Moisture Sensitive
- Moisture Sensitive
- Powder/Lump
Formula:
C₄H₆O₄Sn MW: 236.8 g/mol Melting Pt: 180…182 °C Density: 2.31 g/mL |
MDL Number:
MFCD00054408 CAS Number: 638-39-1 EINECS: 211-335-9 UN: 3146 ADR: 6.1 III |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...