Ethyl-2-bromoisovalerate ≥97%
Supplier: BeanTown Chemical
Synonyms:
2-Bromoisovaleric Acid Ethyl Ester, 2-Bromo-3-methylbutyric Acid Ethyl Ester, Ethyl 2-bromo-3-methylbutanoate, Ethyl 2-Bromo-3-methylbutyrate
136345-25G
136345-5G
BT136345-25GEA
196
USD
BT136345-25G
BT136345-5G
Ethyl-2-bromoisovalerate ≥97%
Ethyl-2-bromoisovalerate
CAS: 609-12-1; EC No: 210-178-3; MDL No: MFCD00026855
UN No: UN3265; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: C7H13BrO2 ; MW: 209.08
Boiling Point: 77°/12 mmHg; Flash point: 65°C (149°F)
Density (g/mL): 1.276
- Liquid
Formula:
C₇H₁₃BrO₂ MW: 209.08 g/mol Boiling Pt: 77 °C/12 mmHg Density: 1.276 g/mL Flash Pt: 65 °C (149 °F) |
MDL Number:
MFCD00026855 CAS Number: 609-12-1 EINECS: 210-178-3 ADR: 8,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...