Iron(II) chloride tetrahydrate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Ferrous chloride tetrahydrate, Iron dichloride tetrahydrate
136335-1KG
136335-250G
BT136335-1KGEA
175.53
USD
BT136335-1KG
BT136335-250G
Iron(II) chloride tetrahydrate ≥98%
Iron(II) chloride tetrahydrate
CAS: 13478-10-9; EC No: 231-843-4; MDL No: MFCD00149709; RTECS: NO5600000
UN No: UN3260; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: FeCl2·4H2O; MW: 198.81
Air Sensitive
- RTECS: NO5600000
- Air Sensitive
- Crystalline
Formula:
FeCl₂·4H₂O MW: 198.81 g/mol |
MDL Number:
MFCD00149709 CAS Number: 13478-10-9 EINECS: 231-843-4 UN: 3260 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...