Silver cyanate ≥98%

Supplier: BeanTown Chemical

136315-100G 136315-25G 136315-5G
BT136315-100GEA 459 USD
BT136315-100G BT136315-25G BT136315-5G
Silver cyanate ≥98%
Silver cyanate

CAS: 3315-16-0; EC No: 222-006-4; MDL No: MFCD00011512 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Linear Formula: AgOCN; MW: 149.89 Density (g/mL): 4 Light Sensitive


  • Light Sensitive
  • Powder

Formula: CAgNO
Density: 4 g/mL
MDL Number: MFCD00011512
CAS Number: 3315-16-0
EINECS: 222-006-4
ADR: 9,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR