N,N-Dimethylformamide ≥99.8% ACS

Supplier: BeanTown Chemical

136300-1L 136300-4L 136300-500ML
BT136300-1LEA 57.77 USD
BT136300-1L BT136300-4L BT136300-500ML
N,N-Dimethylformamide ≥99.8% ACS
N,N-Dimethylformamide

CAS: 68-12-2; EC No: 200-679-5; MDL No: MFCD00003284; RTECS: LQ2100000 UN No: UN2265; Haz Class: 3; Packing Group: III Liquid; Linear Formula: HCON(CH3)2 ; Molecular Formula: C3H7NO; MW: 73.09 Melting Point: -61°; Boiling Point: 153°; Flash point: 57°C (135°F) Density (g/mL): 0.944; Refractive Index: 1.430


  • RTECS: LQ2100000
  • Refractive index: 1.430
  • Liquid

Formula: HCON(CH₃)₂
Boiling Pt: 153 °C
Melting Pt: –61 °C
Density: 0.944 g/mL
Flash Pt: 57 °C (135 °F)
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.8% min.
Color (APHA) ≤10
Residue after evaporation ≤50ppm
Titrable acid ≤0.0005meq/g
Titrable base ≤0.003meq/g
Specific gravity (20/20) Per lot
Methanol Per lot
Iron (Fe) Per lot
Water ≤0.15%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR