1-Naphthol ≥99%
Supplier: BeanTown Chemical
Synonyms:
1-Hydroxynaphthalene, α-Naphthol, α-Hydroxynaphthalene
136290-100G
136290-2.5KG
136290-500G
BT136290-100GEA
36.57
USD
BT136290-100G
BT136290-2.5KG
BT136290-500G
1-Naphthol ≥99%
1-Naphthol
CAS: 90-15-3; EC No: 201-969-4; MDL No: MFCD00003930; RTECS: QL2800000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C10H8O; MW: 144.17
Melting Point: 94-96°; Boiling Point: 278-280°; Flash point: 125°C (257°F)
Air Sensitive, Light Sensitive
- RTECS: QL2800000
- Air Sensitive, Light Sensitive
- Solid
Formula:
C₁₀H₈O MW: 144.17 g/mol Boiling Pt: 278…280 °C Melting Pt: 94…96 °C Flash Pt: 125 °C (257 °F) |
MDL Number:
MFCD00003930 CAS Number: 90-15-3 EINECS: 201-969-4 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...