4-Fluoro-o-phenylenediamine ≥97%

Supplier: BeanTown Chemical

Synonyms: 3,4-Diamino-1-fluorobenzene, 4-Fluorobenzene-1,2-diamine, 1,2-Diamino-4-fluorobenzene, 4-Fluoro-1,2-phenylenediamine

136285-100G 136285-25G 136285-5G
BT136285-100GEA 1039 USD
BT136285-100G BT136285-25G BT136285-5G
4-Fluoro-o-phenylenediamine ≥97%
4-Fluoro-o-phenylenediamine

CAS: 367-31-7; EC No: 206-691-7; MDL No: MFCD00042228 UN No: UN2811; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: C6H7FN2; MW: 126.13 Melting Point: 94-98° Light Sensitive


  • Light Sensitive
  • Powder

Formula: C₆H₇FN₂
Melting Pt: 94…98 °C
MDL Number: MFCD00042228
CAS Number: 367-31-7
EINECS: 206-691-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR