α-Bromo-4-iodotoulene ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-Iodobenzyl bromide, 1-(Bromomethyl)-4-iodobenzene, α-Bromo-p-iodotoulene

136230-100G 136230-25G 136230-5G
BT136230-100GEA 1124 USD
BT136230-100G BT136230-25G BT136230-5G
α-Bromo-4-iodotoulene ≥97%
α-Bromo-4-iodotoulene

CAS: 16004-15-2; MDL No: MFCD00209656 UN No: UN3261; Haz Class: 8; Packing Group: II Crystalline; Molecular Formula: C7H6BrI; MW: 296.93 Melting Point: 78-82° Light Sensitive


  • Light Sensitive
  • Crystalline

Formula: C₇H₆BrI
MW: 296.93
Melting Pt: 80
MDL Number: MFCD00209656
CAS Number: 16004-15-2
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR