Triisopropylphosphine ≥98%

Supplier: BeanTown Chemical

136190-10G 136190-2G 136190-500MG
BT136190-10GEA 881 USD
BT136190-10G BT136190-2G BT136190-500MG
Triisopropylphosphine ≥98%
Triisopropylphosphine

CAS: 6476-36-4; EC No: 229-333-1; MDL No: MFCD00015029 UN No: UN2845; Haz Class: 4.2; Packing Group: I Liquid; Linear Formula: [(CH3)2CH]3P; Molecular Formula: C9H21P; MW: 160.24 Boiling Point: 176-178°; Flash point: 105°C (221°F) Density (g/mL): 0.839 ; Refractive Index: 1.466 Air Sensitive


  • Refractive index: 1.466
  • Air Sensitive
  • Liquid

Formula: C₉H₂₁P
Boiling Pt: 176…178 °C
Density: 0.839 g/mL
Flash Pt: 105 °C (221 °F)
MDL Number: MFCD00015029
CAS Number: 6476-36-4
EINECS: 229-333-1
UN: 2845
ADR: 4.2,I

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR