4,4'-Bipyridyl ≥99%
Supplier: BeanTown Chemical
Synonyms:
4,4'-Bipyridine, 4,4'-Dipyridyl
136180-100G
136180-25G
136180-5G
BT136180-100GEA
313
USD
BT136180-100G
BT136180-25G
BT136180-5G
4,4'-Bipyridyl ≥99%
4,4'-Bipyridyl
CAS: 553-26-4; EC No: 209-036-3; MDL No: MFCD00006416; RTECS: DW1760000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: C10H8N2; MW: 156.18
Melting Point: 109-112°; Boiling Point: 305°
Hygroscopic
- RTECS: DW1760000
- Hygroscopic
- Powder
Formula:
C₁₀H₈N₂ MW: 156.19 g/mol Boiling Pt: 305 °C Melting Pt: 109…112 °C |
MDL Number:
MFCD00006416 CAS Number: 553-26-4 EINECS: 209-036-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...