Dibutyltin oxide ≥98%

Supplier: BeanTown Chemical

Synonyms: Dibutyltin(IV) oxide, Dibutyloxostannane, Di-n-butyltin oxide, Di-n-butyltin(IV) oxide

136090-100G 136090-25G 136090-500G
BT136090-100GEA 37 USD
BT136090-100G BT136090-25G BT136090-500G
Dibutyltin oxide ≥98%
Dibutyltin oxide

CAS: 818-08-6; EC No: 212-449-1; MDL No: MFCD00001992; RTECS: WH7175000 UN No: UN3146; Haz Class: 6.1; Packing Group: II Powder; Linear Formula: (CH3CH2CH2CH2)2SnO; Molecular Formula: C8H18OSn; MW: 248.92 Melting Point: 300°


  • RTECS: WH7175000
  • Powder

Formula: C₈H₁₈OSn
MW: 248.94 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00001992
CAS Number: 818-08-6
EINECS: 212-449-1
UN: 3146
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR