n-Heptane ≥99.0% for HPLC
Supplier: BeanTown Chemical
Synonyms:
high purity solvents
136075-1L
136075-250ML
BT136075-1LEA
69.83
USD
BT136075-1L
BT136075-250ML
n-Heptane ≥99.0% for HPLC
n-Heptane
CAS: 142-82-5; EC No: 205-563-8; MDL No: MFCD00009544; RTECS: MI7700000
UN No: UN1206; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3(CH2)5CH3; Molecular Formula: C7H16 ; MW: 100.21
Melting Point: -91°; Boiling Point: 97-99°; Flash point: -4°C (24°F)
Density (g/mL): 0.684; Refractive Index: 1.387
- RTECS: MI7700000
- Refractive index: 1.387
- Liquid
Formula:
H₃C(CH₂)₅CH₃ MW: 100.20 g/mol Boiling Pt: 98.4 Melting Pt: -90.6 Density: 0.684 g/mL Flash Pt: –4 °C (24 °F) |
MDL Number:
MFCD00009544 CAS Number: 142-82-5 EINECS: 205-563-8 UN: 1206 ADR: 3 II Merck Index: 13,04679 |
|
Specification Test Results
Assay | 99.0% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...