Ruthenium(III) nitrosylnitrate 1,5% (w/v) in water

Supplier: BeanTown Chemical

136070-1L 136070-250ML 136070-50ML
BT136070-1LEA 1387 USD
BT136070-1L BT136070-250ML BT136070-50ML
Ruthenium(III) nitrosylnitrate 1,5% (w/v) in water
Ruthenium(III) nitrosylnitrate

CAS: 34513-98-9; EC No: 252-068-8; MDL No: MFCD00016313 UN No: UN3218; Haz Class: 5.1; Packing Group: III Liquid; Molecular Formula: N4O10Ru; MW: 317.09 Density (g/mL): 1.1


  • Liquid

Formula: N₄O₁₀Ru
MW: 317.09 g/mol
Density: 1.1 g/mL
MDL Number: MFCD00016313
CAS Number: 34513-98-9
EINECS: 252-068-8
ADR: 5.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR