4-Bromothiophenol ≥98%

Supplier: BeanTown Chemical

135990-10G 135990-250G 135990-50G
BT135990-10GEA 53.33 USD
BT135990-10G BT135990-250G BT135990-50G
4-Bromothiophenol ≥98%
4-Bromothiophenol

CAS: 106-53-6; EC No: 203-407-3; MDL No: MFCD00004845 UN No: UN2923; Haz Class: 8 (6.1); Packing Group: III Crystalline; Molecular Formula: C6H5BrS; MW: 189.07 Melting Point: 73-76°; Boiling Point: 239° Air Sensitive


  • Air Sensitive
  • Crystalline

Formula: C₆H₅BrS
Boiling Pt: 239 °C
Melting Pt: 73…76 °C
MDL Number: MFCD00004845
CAS Number: 106-53-6
EINECS: 203-407-3
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR