Diethyl sulfide ≥96%

Supplier: BeanTown Chemical

135975-10G 135975-50G
BT135975-10GEA 40.07 USD
BT135975-10G BT135975-50G
Diethyl sulfide ≥96%
Diethyl sulphide

CAS: 352-93-2; EC No: 206-526-9; MDL No: MFCD00009270; RTECS: LC7200000 UN No: UN2375; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3CH2SCH2CH3; Molecular Formula: C4H10S; MW: 90.19 Melting Point: -100°; Boiling Point: 90-92°; Flash point: -10°C (14°F) Density (g/mL): 0.837; Refractive Index: 1.442


  • RTECS: LC7200000
  • Refractive index: 1.442
  • Liquid

Formula: C₄H₁₀S
MW: 90.19 g/mol
Boiling Pt: 90…92 °C
Melting Pt: –100 °C
Density: 0.837 g/mL
Flash Pt: –10 °C (14 °F)
MDL Number: MFCD00009270
CAS Number: 352-93-2
EINECS: 206-526-9
UN: 2375
ADR: 3,II

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR