3,4-Diaminoanisole ≥98%
Supplier: BeanTown Chemical
Synonyms:
4-Methoxy-o-phenylenediamine
135945-1G
135945-25G
135945-5G
BT135945-1GEA
40
USD
BT135945-1G
BT135945-25G
BT135945-5G
3,4-Diaminoanisole ≥98%
3,4-Diaminoanisole
CAS: 102-51-2; MDL No: MFCD00047837; RTECS: ST2690000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C7H10N2O; MW: 138.17
Melting Point: 46-50°
Air Sensitive, Light Sensitive
- RTECS: ST2690000
- Air Sensitive, Light Sensitive
- Solid
Formula:
C₇H₁₀N₂O MW: 138.17 g/mol Melting Pt: 46…50 °C |
MDL Number:
MFCD00047837 CAS Number: 102-51-2 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...