2,4,6-Trimethylaniline ≥98%
Supplier: BeanTown Chemical
CAS: 88-05-1; EC No: 201-794-3; MDL No: MFCD00007740; RTECS: BZ0700000
UN No: UN2810; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C9H13N; MW: 135.21
Boiling Point: 233°; Flash point: 96°C (205°F)
Density (g/mL): 0.963; Refractive Index: 1.551
- RTECS: BZ0700000
- Refractive index: 1.551
- Liquid
Formula:
C₉H₁₃N Boiling Pt: 233 °C Density: 0.963 g/mL Flash Pt: 96 °C (205 °F) |
MDL Number:
MFCD00007740 CAS Number: 88-05-1 EINECS: 201-794-3 UN: 2810 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...