Isoamyl alcohol ≥98.5% ACS

Supplier: BeanTown Chemical

Synonyms: Isopentyl alcohol, iso-Pentanol, 3-Methyl-1-butanol, 3-Methylbutanol

135835-1L 135835-4L 135835-500ML
BT135835-1LEA 83.35 USD
BT135835-1L BT135835-4L BT135835-500ML
Isoamyl alcohol ≥98.5% ACS
Isoamyl alcohol

CAS: 123-51-3; EC No: 204-633-5; MDL No: MFCD00002934; RTECS: EL5425000 UN No: UN1105; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)2CHCH2CH2OH; Molecular Formula: C5H12O; MW: 88.15 Melting Point: -117°; Boiling Point: 130°; Flash point: 43°C (109°F) Density (g/mL): 0.809; Refractive Index: 1.406


  • RTECS: EL5425000
  • Refractive index: 1.406
  • Liquid

Formula: (CH₃)₂CHCH₂CH₂OH
MW: 88.15 g/mol
Boiling Pt: 130 °C
Melting Pt: –117 °C
Density: 0.809 g/mL
Flash Pt: 43 °C (109 °F)
MDL Number: MFCD00002934
CAS Number: 123-51-3
EINECS: 204-633-5
UN: 1105
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=170c60e26c7b4edab6e0686f91d04e676da16e6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98.5% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR