Barium hydroxide, anhydrous ≥95%

Supplier: BeanTown Chemical

135820-1KG 135820-250G 135820-50G
BT135820-1KGEA 103.32 USD
BT135820-1KG BT135820-250G BT135820-50G
Barium hydroxide, anhydrous ≥95%
Barium hydroxide

CAS: 17194-00-2; EC No: 241-234-5; MDL No: MFCD00003443 UN No: UN3262; Haz Class: 8; Packing Group: III Powder; Molecular Formula: H2BaO2 ; MW: 171.34 Melting Point: 300° Density (g/mL): 2.2 Air Sensitive , Hygroscopic


  • Air Sensitive, Hygroscopic
  • Powder

Formula: H₂BaO₂
Melting Pt: >300 °C
Density: 2.2 g/mL
MDL Number: MFCD00003443
CAS Number: 17194-00-2
EINECS: 241-234-5
UN: 3262
ADR: 8,III

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR