2,6-Dichloronicotinonitrile ≥97%
Supplier: BeanTown Chemical
Synonyms:
2,6-Dichloropyridine-3-carbonitrile, 2,6-Dichloro-3-cyanopyridine
135770-1G
135770-250MG
135770-5G
BT135770-1GEA
89
USD
BT135770-1G
BT135770-250MG
BT135770-5G
2,6-Dichloronicotinonitrile ≥97%
2,6-Dichloronicotinonitrile
CAS: 40381-90-6; MDL No: MFCD03411341
UN No: UN2811 ; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: C6H2Cl2N2; MW: 173.00
Melting Point: 121-125°
Moisture Sensitive
- Moisture Sensitive
- Powder
Formula:
C₆H₂Cl₂N₂ MW: 173.00 g/mol Melting Pt: 121…125 °C |
MDL Number:
MFCD03411341 CAS Number: 40381-90-6 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...