2,5-Dimethylhexane ≥99%

Supplier: BeanTown Chemical

135760-25G 135760-5G
BT135760-25GEA 299 USD
BT135760-25G BT135760-5G
2,5-Dimethylhexane ≥99%
2,5-Dimethylhexane

CAS: 592-13-2; EC No: 209-745-8; MDL No: MFCD00008951 UN No: UN3295; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)2CHCH2CH2CH(CH3)2; Molecular Formula: C8H18; MW: 114.23 Boiling Point: 108°; Flash point: 26°C (79°F) Density (g/mL): 0.694; Refractive Index: 1.392


  • Refractive index: 1.392
  • Liquid

Formula: C₈H₁₈
MW: 114.23 g/mol
Boiling Pt: 108 °C
Density: 0.694 g/mL
Flash Pt: 26 °C (79 °F)
MDL Number: MFCD00008951
CAS Number: 592-13-2
EINECS: 209-745-8
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR