Trifluoroacetic acid-[D₁] ≥99.5% (Isotopic)

Supplier: BeanTown Chemical

135660-10G 135660-25G
BT135660-10GEA 55 USD
BT135660-10G BT135660-25G
Trifluoroacetic acid-[D₁] ≥99.5% (Isotopic)
Trifluoroacetic acid-[D₁]

CAS: 599-00-8; EC No: 209-961-2; MDL No: MFCD00044563 UN No: UN2699; Haz Class: 8; Packing Group: I Liquid, in prescored ampules; Linear Formula: CF3COOD; MW: 115.03 Melting Point: -15°; Boiling Point: 75° Density (g/mL): 1.493; Refractive Index: 1.284 Hygroscopic


  • Refractive index: 1.284
  • Hygroscopic
  • Liquid, in prescored ampules

Formula: F₃CCOOD
MW: 115.02 g/mol
Boiling Pt: 75 °C
Melting Pt: –15 °C
Density: 1.493 g/mL
MDL Number: MFCD00044563
CAS Number: 599-00-8
EINECS: 209-961-2
UN: 2699
ADR: 8,I

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR