Benzaldehyde ≥99%
Supplier: BeanTown Chemical
135650-1KG
135650-250G
135650-5KG
BT135650-1KGEA
58.57
USD
BT135650-1KG
BT135650-250G
BT135650-5KG
Benzaldehyde ≥99%
Benzaldehyde
CAS: 100-52-7; EC No: 202-860-4; MDL No: MFCD00003299; RTECS: CU4375000
UN No: UN1990; Haz Class: 9; Packing Group: III
Liquid; Molecular Formula: C7H6O; MW: 106.12
Melting Point: -26°; Boiling Point: 178-179°; Flash point: 62°C (143°F)
Density (g/mL): 1.044; Refractive Index: 1.545
Air Sensitive
- RTECS: CU4375000
- Refractive index: 1.545
- Air Sensitive
- Liquid
Formula:
C₆H₅CHO MW: 106.12 g/mol Boiling Pt: 178…179 °C Melting Pt: –26 °C Density: 1.044 g/mL Flash Pt: 62 °C (143 °F) |
MDL Number:
MFCD00003299 CAS Number: 100-52-7 EINECS: 202-860-4 UN: 1990 ADR: 9,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...