1-Chloro-3,3-dimethylbutane ≥97%
Supplier: BeanTown Chemical
135640-10G
135640-50G
BT135640-10GEA
84.02
USD
BT135640-10G
BT135640-50G
1-Chloro-3,3-dimethylbutane ≥97%
1-Chloro-3,3-dimethylbutane
CAS: 2855-08-5; EC No: 220-665-2; MDL No: MFCD00018980
UN No: UN1993; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: (CH3)3CCH2CH2Cl; Molecular Formula: C6H13Cl; MW: 120.62
Flash point: 7°C (44°F)
Density (g/mL): 0.87
- Liquid
Formula:
C₆H₁₃Cl Density: 0.87 g/mL Flash Pt: 7 °C (44 °F) |
MDL Number:
MFCD00018980 CAS Number: 2855-08-5 EINECS: 220-665-2 ADR: 3,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...