Bismuth(III) nitrate, hydrate ≥99.999% (trace metals basis)
Supplier: BeanTown Chemical
|
Danger
|
Synonyms:
Bismuth trinitrate
135625-1G
135625-25G
135625-5G
BT135625-1GEA
64
USD
BT135625-1G
BT135625-25G
BT135625-5G
Bismuth(III) nitrate, hydrate ≥99.999% (trace metals basis)
Bismuth(III) nitrate
CAS: 10361-44-1; EC No: 233-791-8; MDL No: MFCD00003460
UN No: UN1477; Haz Class: 5.1; Packing Group: II
Crystalline; Linear Formula: Bi(NO3)3·xH2O; MW: 394.99 (anhydrous)
Melting Point: 30°; Boiling Point: 75-80°
Hygroscopic
Formula:
Bi(NO₃)₃·xH₂O MW: 395 Boiling Pt: 75…80 °C Melting Pt: 845 Density: 2.83 |
MDL Number:
MFCD00003460 CAS Number: 10361-44-1 EINECS: 233-791-8 UN: 1477 ADR: 5.1 II Merck Index: 14,1271 |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...