Copper(II) nitrate hemipentahydrate ≥98%
Supplier: BeanTown Chemical
135605-1KG
135605-250G
135605-5KG
BT135605-1KGEA
122.45
USD
BT135605-1KG
BT135605-250G
BT135605-5KG
Copper(II) nitrate hemipentahydrate ≥98%
Copper(II) nitrate hemipentahydrate
CAS: 19004-19-4; EC No: 221-838-5; MDL No: MFCD00149671; RTECS: GL7875000
UN No: UN3085; Haz Class: 5.1 (8); Packing Group: III
Solid; Linear Formula: Cu(NO3)2·2.5H2O; MW: 232.59
Density (g/mL): 2.32
Hygroscopic
- RTECS: GL7875000
- Hygroscopic
- Solid
Formula:
Cu(NO₃)₂·2.5H₂O Density: 2.32 g/mL |
MDL Number:
MFCD02253001 CAS Number: 19004-19-4 EINECS: 221-838-5 UN: 3085 ADR: 5.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...