Gallium trinitrate hydrate ≥99.9%
Supplier: BeanTown Chemical
135555-25G
135555-500MG
135555-5G
BT135555-25GEA
226.06
USD
BT135555-25G
BT135555-500MG
BT135555-5G
Gallium trinitrate hydrate ≥99.9%
Gallium trinitrate hydrate
CAS: 69365-72-6; EC No: 236-815-5; MDL No: MFCD00149723
UN No: UN1477; Haz Class: 5.1; Packing Group: III
Lump; Linear Formula: Ga(NO3)3·xH2O; MW: 255.73 (anhydrous)
Melting Point: 110° (decomposes)
Hygroscopic
- Hygroscopic
- Lump
Formula:
GaN₃O₉·H₂O MW: 273.75 Melting Pt: 110 °C (dec.) |
MDL Number:
MFCD00149723 CAS Number: 69365-72-6 EINECS: 236-815-5 UN: 1477 ADR: 5.1 III |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...