4-Nitro-2-picoline-N-oxide ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Methyl-4-nitropyridine-1-oxide, 4-Nitro-2-picoline N-oxide, 2-Methyl-4-nitropyridine-N-oxide
135500-1G
135500-25G
135500-5G
BT135500-1GEA
34
USD
BT135500-1G
BT135500-25G
BT135500-5G
4-Nitro-2-picoline-N-oxide ≥98%
4-Nitro-2-picoline-N-oxide
CAS: 5470-66-6; MDL No: MFCD00014625; RTECS: UT5710000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C6H7N2O3; MW: 154.12
Melting Point: 155-159°
- RTECS: UT5710000
- Solid
Formula:
C₆H₆N₂O₃ MW: 154.13 g/mol Melting Pt: 156 |
MDL Number:
MFCD00014625 CAS Number: 5470-66-6 ADR: 6.1 III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...