Bis(cyclopentadienyl)zirconium hydridochloride ≥95%

Supplier: BeanTown Chemical

Synonyms: Bis(cyclopentadienyl)zirconium(IV) chloride hydride, Schwartz's Reagent, Bis(cyclopentadienyl)zirconium chloride hydride

135415-1G 135415-25G 135415-5G
BT135415-1GEA 41 USD
BT135415-1G BT135415-25G BT135415-5G
Bis(cyclopentadienyl)zirconium hydridochloride ≥95%
Bis(cyclopentadienyl)zirconium hydridochloride

CAS: 37342-97-5; EC No: 253-479-5; MDL No: MFCD02089401 UN No: UN3131; Haz Class: 4.3 (8); Packing Group: II Solid; Molecular Formula: C10H11ClZr; MW: 257.87 Air Sensitive, Light Sensitive, Moisture Sensitive


  • Air Sensitive, Light Sensitive, Moisture Sensitive
  • Solid

Formula: C₁₀H₁₁ClZr
MW: 257.87 g/mol
MDL Number: MFCD02089401
CAS Number: 37342-97-5
EINECS: 253-479-5
UN: 3131
ADR: 4.3 II

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR