Nickel sulfide ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Nickel subsulphide, Nickel disulphane

135400-10G 135400-250G 135400-50G
BT135400-10GEA 83 USD
BT135400-10G BT135400-250G BT135400-50G
Nickel sulfide ≥99.9%
Nickel sulfide

CAS: 12035-72-2; EC No: 234-829-6; MDL No: MFCD00016266; RTECS: QR9800000 UN No: UN3077; Haz Class: 9; Packing Group: III -200 Mesh Powder; Molecular Formula: Ni3S2; MW: 240.20 Melting Point: 797° Density (g/mL): 5.8


  • RTECS: QR9800000
  • -200 Mesh Powder

Formula: Ni₃S₂
Melting Pt: 797 °C
Density: 5.8 g/mL
MDL Number: MFCD00217044
CAS Number: 12035-72-2
EINECS: 234-829-6
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR