Zinc acetate, anhydrous ≥99.95% (trace metals basis)
Supplier: BeanTown Chemical
135385-10G
135385-50G
BT135385-10GEA
58.57
USD
BT135385-10G
BT135385-50G
Zinc acetate, anhydrous ≥99.95% (trace metals basis)
Zinc acetate
CAS: 557-34-6; EC No: 209-170-2; MDL No: MFCD00012454; RTECS: AK1500000
UN No: UN3077; Haz Class: 9; Packing Group: III
Granular; Linear Formula: (CH3CO2)2Zn; MW: 183.47
Melting Point: 200° (decomposes)
Density (g/mL): 1.84
Hygroscopic
- RTECS: AK1500000
- Hygroscopic
- Granular
Formula:
C₄H₆O₄Zn MW: 183.48 g/mol Melting Pt: 200 °C (dec.) Density: 1.84 g/mL |
MDL Number:
MFCD00012454 CAS Number: 557-34-6 EINECS: 209-170-2 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 99.95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...