1,1,3,3-Tetramethoxypropane ≥98%

Supplier: BeanTown Chemical

135355-1KG 135355-250G 135355-5KG
BT135355-1KGEA 124 USD
BT135355-1KG BT135355-250G BT135355-5KG
1,1,3,3-Tetramethoxypropane ≥98%
1,1,3,3-Tetramethoxypropane

CAS: 102-52-3; EC No: 203-037-2; MDL No: MFCD00008488 UN No: UN3271; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C7H16O4 ; MW: 164.20 Boiling Point: 183°; Flash point: 60°C (140°F) Density (g/mL): 0.997; Refractive Index: 1.407


  • Refractive index: 1.407
  • Liquid

Formula: C₇H₁₆O₄
Boiling Pt: 183 °C
Density: 0.997 g/mL
Flash Pt: 60 °C (140 °F)
MDL Number: MFCD00008488
CAS Number: 102-52-3
EINECS: 203-037-2
UN: 3271
ADR: 3,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR