α-Bromo-2-nitrotoluene ≥98%
Supplier: BeanTown Chemical
Synonyms:
α-Bromo-o-nitrotoluene, 1-(Bromomethyl)-2-nitrobenzene, alpha-Bromo-2-nitrotoluene, 2-Nitrobenzyl bromide
135300-100G
135300-25G
135300-5G
BT135300-100GEA
647
USD
BT135300-100G
BT135300-25G
BT135300-5G
α-Bromo-2-nitrotoluene ≥98%
α-Bromo-2-nitrotoluene
CAS: 3958-60-9; EC No: 223-558-9; MDL No: MFCD00007184
UN No: UN3261; Haz Class: 8; Packing Group: II
Crystalline; Molecular Formula: C7H6BrNO2; MW: 216.03
Melting Point: 44-46°; Flash point: 110°C (230°F)
- Crystalline
Formula:
C₇H₆BrNO₂ MW: 216.03 g/mol Melting Pt: 44…46 °C Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00007184 CAS Number: 3958-60-9 EINECS: 223-558-9 UN: 3261 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...