Niobium ≥99.5% (trace metals basis), Powder -325 mesh

Supplier: BeanTown Chemical

135285-100G 135285-25G
BT135285-100GEA 167.08 USD
BT135285-100G BT135285-25G
Niobium ≥99.5% (trace metals basis), Powder -325 mesh
Niobium

CAS: 7440-03-1; EC No: 231-113-5; MDL No: MFCD00011126; RTECS: QT9900000 UN No: UN3089; Haz Class: 4.1; Packing Group: II Powder; Molecular Formula: Nb ; MW: 92.91 Melting Point: 2468°; Boiling Point: 4742° Density (g/mL): 8.57


  • RTECS: QT9900000
  • Powder

Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 4742 °C
Melting Pt: 2468 °C
Density: 8.57 g/mL
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5
UN: 3089
ADR: 4.1,II

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR