3-Methyl-2-butenal ≥97%

Supplier: BeanTown Chemical

Synonyms: 3,3-Dimethylacrolein, 3-Methylbut-2-enal, Senecialdehyde, 3-Methylcrotonaldehyde

135240-100G 135240-25G 135240-5G
BT135240-100GEA 630 USD
BT135240-100G BT135240-25G BT135240-5G
3-Methyl-2-butenal ≥97%
3-Methyl-2-butenal

CAS: 107-86-8; EC No: 203-527-6; MDL No: MFCD00010291 UN No: UN2929; Haz Class: 6.1 (3); Packing Group: II Liquid; Molecular Formula: C5H8O; MW: 84.12 Boiling Point: 133-135°; Flash point: 33°C (91°F) Density (g/mL): 0.872; Refractive Index: 1.462 Air Sensitive


  • Refractive index: 1.462
  • Air Sensitive
  • Liquid

Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 133…135 °C
Density: 0.872 g/mL
Flash Pt: 33 °C (91 °F)
MDL Number: MFCD00010291
CAS Number: 107-86-8
EINECS: 203-527-6
ADR: 6.1 (3),II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR