Trichloroacetyl chloride ≥99%

Supplier: BeanTown Chemical

135235-1KG 135235-250G 135235-50G
BT135235-1KGEA 784 USD
BT135235-1KG BT135235-250G BT135235-50G
Trichloroacetyl chloride ≥99%
Trichloroacetyl chloride

CAS: 76-02-8; EC No: 200-926-7; MDL No: MFCD00000792; RTECS: AO7140000 UN No: UN2442; Haz Class: 8 (6.1); Packing Group: II Liquid; Linear Formula: Cl3CCOCl; Molecular Formula: C2Cl4O; MW: 181.83 Melting Point: -57°; Boiling Point: 114-116°; Flash point: 100°C (212°F) Density (g/mL): 1.629; Refractive Index: 1.470 Moisture Sensitive


  • RTECS: AO7140000
  • Refractive index: 1.470
  • Moisture Sensitive
  • Liquid

Formula: C₂Cl₄O
MW: 181.83 g/mol
Boiling Pt: 114…116 °C
Melting Pt: –57 °C
Density: 1.629 g/mL
Flash Pt: 100 °C (212 °F)
MDL Number: MFCD00000792
CAS Number: 76-02-8
EINECS: 200-926-7
UN: 2442
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR