Suxamethonium chloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Succinylcholine chloride
135200-1G
135200-5G
BT135200-1GEA
31
USD
BT135200-1G
BT135200-5G
Suxamethonium chloride ≥98%
Suxamethonium chloride
CAS: 71-27-2; EC No: 200-747-4; MDL No: MFCD00038725; RTECS: GA2360000
UN No: UN3249; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: C14H30Cl2N2O4; MW: 361.30
- RTECS: GA2360000
- Powder
Formula:
C₁₄H₃₀Cl₂N₂O₄ |
MDL Number:
MFCD00038725 CAS Number: 71-27-2 EINECS: 200-747-4 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...